Ferrules, Alumaseal®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21472 21473 21474
REST21472EA 115 EUR
REST21472 REST21473 REST21474
Ferrules, Alumaseal®
Fittingen en connectoren
These ferrules are made of aluminium eliminate out-gassing and make leak-tight seals for less detector noise. Its unique two-piece design permanently locks on fused silica tubing without causing breakage.
  • No retightening required after temperature cycles
  • Will not crack or fragment
  • Will not stick in fittings
  • Use with any 1,6 mm compression-type fitting
  • Stable up to 550 °C