Oven temperature sensor assembly for Agilent GCs
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
23039 23040
REST23039EA 158 EUR
REST23039 REST23040
Oven temperature sensor assembly for Agilent GCs
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The oven sensors works in conjunction with the oven control board to regulate oven temperature.
For use with Agilent 5890 and 6890 GCs.