Hazardous substance collection stations, HazMat
Leverancier: CEMO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3412 5229
CEMG3412EA 2390 EUR
CEMG3412 CEMG5229
Hazardous substance collection stations, HazMat
Opslag voor vaten
Hazardous substance collection stations, made of glass fibre reinforced plastic (GRP), offer central, safe storage for different containers and liquids. The GRP sump pallet with splash protection wall is suitable to store liquids of all the waterpolluting classes.
  • With galvanised floor frame with grating
  • Protected by patent P 38 01 810.1-09
  • Allows outdoor or indoor use
  • With 200 l containment space
  • Wide opening, lockable lid with pneumatic springs enables easy loading/unloading
  • With general technical approval no. Z-40.12-228 granted by DIBt-Berlin
Surface area: 1500×850 mm (L×W)