Condenser, air, CondenSyn™
Leverancier: ASYNT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GB-C-350-B19 GB-C-350-B29 GB-C-450-B29 GB-C-200-B29 GB-C-350-B24 GB-C-450-B19 GB-C-450-B24 GB-C-350-B14 GB-C-450-B14 GB-C-200-B14 GB-C-200-B19 GB-C-200-B24
ASYNGB-C-350-B19EA 417 EUR
ASYNGB-C-350-B19 ASYNGB-C-350-B29 ASYNGB-C-450-B29 ASYNGB-C-200-B29 ASYNGB-C-350-B24 ASYNGB-C-450-B19 ASYNGB-C-450-B24 ASYNGB-C-350-B14 ASYNGB-C-450-B14 ASYNGB-C-200-B14 ASYNGB-C-200-B19 ASYNGB-C-200-B24
Condenser, air, CondenSyn™
Condensors
Condenser made of borosilicate glass.
  • Robust and easy to clean
  • Suitable for use under vacuum
  • High performance and environmentally friendly
  • No risk of flooding
By not using water the environmental impact is reduced as is the costs of water usage.
200 mm recommended for use with flasks up to 100 mL in size (Not more than half full).
350 mm recommended for use with flasks up to 500 mL in size (Not more than half full).
450 mm recommended for use with flasks up to 1000 mL in size (Not more than half full).