Miniature passive air samplers
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26252 26253 26254 26255 26256 26257 26258 26259
REST26252EA 1450 EUR
REST26252 REST26253 REST26254 REST26255 REST26256 REST26257 REST26258 REST26259
Miniature passive air samplers
Monsternemers Luchtmonsternametoestellen
These air samplers provide accurate integrated sampling via small fittings that easily connect to miniature canisters.
Bestelinformatie: Miniature canisters are not included and must be order separately.
Leveringsinformatie: Includes a flow controller, critical orifice, vaccum gauge, sample inlet, and frit filter.