FID capillary column adapters
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22608 22609
REST22608EA 133 EUR
REST22608 REST22609
FID capillary column adapters
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These FID capillary column adapters made of stainless steel are ideal for PerkinElmer Auto SYS XL.