Tubing, stainless steel, straight
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22902 22901
REST22902EA 663 EUR
REST22902 REST22901
Tubing, stainless steel, straight
Slangen Transfer Tubing
These stainless steel tubing has individual 1,83 m length straight pieces. This tubing can be cut to exact requirements using a standard tubing cutter.