Passive air samplers
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
24160 24161 24162 24163 24164 24165 24166 24167 24168 24169 24216 24217
REST24160EA 1750 EUR
REST24160 REST24161 REST24162 REST24163 REST24164 REST24165 REST24166 REST24167 REST24168 REST24169 REST24216 REST24217
Passive air samplers
Monsternemers Luchtmonsternametoestellen
These passive air samplers provide accurate integrated sampling without a sampling pump.
  • Siltek® treated components ensure accurate sampling of active components
  • Excellent for sampling times from 0,5 to 125 hours
Bestelinformatie: Flow controllers and critical orifices can be ordered separately.
Leveringsinformatie: Includes Veriflo® SC423XL flow controller, stainless steel vaccum gauge, siltek® sample inlet of 6,35 mm (1/4"), 2 µm frit filter with washer and interchangeable critical orifice.