Silver seals
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
23057 23058
REST23057EA 179 EUR
REST23057 REST23058
Silver seals
Afdichtingen Mechanische afdichtingen
Silver seals for Thermo Scientific TRACE, 8000, 8000 TOP and Focus.