Claw and claw holder kit
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26261 26262
REST26261EA 53 EUR
REST26261 REST26262
Claw and claw holder kit
Uittrek- en invoeggereedschap
This claw safely and cleanly removes liners, O-rings, or other small objects from the injection port.
The hot objects can be placed in a clean 4 ml vial situated in the claw holder until ready for reuse.