Fittings, brass and stainless steel, Parker®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21800 I21801 21818 21832 21835 21837 21841 21844 21847 21908 21929 21930 21932 21941 23187 21948 21958 21960 21961
REST21800EA 0 EUR
REST21800 JJBEI21801 REST21818 REST21832 REST21835 REST21837 REST21841 REST21844 REST21847 REST21908 REST21929 REST21930 REST21932 REST21941 REST23187 REST21948 REST21958 REST21960 REST21961
Fittings, brass and stainless steel, Parker®
Fittingen en connectoren
Parker® (A-Lok) fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections. Fittings are available in brass or 316 grade stainless steel.