Toray carbon paper, TGP-H-60
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45365.KS 45356.KS
45365.KSEA 171 EUR
45365.KS 45356.KS
Toray carbon paper, TGP-H-60
Papier
Toray carbon paper, TGP-H-60 can be used as a gas diffusion layer in fuel cell applications. It is also suitable for use as a catalyst backing layer.
  • Insoluble in water
  • Air and light sensitive
  • Container must be closed tightly and stored in a cool, dry and well-ventilated place
It is incompatible with strong oxidizing agents, chlorinated paraffins, lead oxide, manganese oxide, iron oxide, liquid oxygen, oils, chlorinetrifluoride, fluoride and potassium superoxide.
Bestelinformatie: Available in PTFE treated version.