Koelthermostaten, serie Alpha
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L000643 L000644 L000645 L000646 L000638 L000639 L000640 L000641 L000642
LAUDL000643EA 4370 EUR
LAUDL000643 LAUDL000644 LAUDL000645 LAUDL000646 461-1249 461-1250 461-1251 LAUDL000641 LAUDL000642
Koelthermostaten, serie Alpha
Circulatiethermostaten Gesloten circulatiesystemen Koelingscirculatoren
The Alpha cooling thermostats have an easy to use control console featuring three button operation and large, clear LED display, ensuring easy navigation through the menu. The thermostats are suitable for use with non flammable liquids (water, water/glycol mixtures). Units can provide both internal and external thermostating. Temperature stability of ±0,05 K gives precise heating and cooling suitable for a wide range of applications. The automatic cooling system enables cooling throughout the entire temperature range of −25 to +100 °C. All cooling thermostats contain a compressor system which can be switched on or off as required, thus conserving energy and saving wear and tear on the compressor to give a longer working life.
  • Temperature range −25 to +100 °C, resolution 0,1 °C, temperature stability of ±0,05 K
  • User calibration at a single point for re-calibration of working temperatures
  • Safety functions such as alarms, warnings and errors are shown on the LED display
  • The front plate can be quickly removed to make cleaning the condenser easy, a drain connection at the rear of the unit makes emptying the temperature fluid simple
  • Units have an integrated timer which automatically shuts down to standby mode after a preset time
Maximum pump capacities:
15 L/min, 200 mbar.
15 l/min, 200 mbar
Leveringsinformatie: Geleverd met baddeksels en pompkits met pompaansluitingen van Ø 13 mm.
Opgelet: Cooling capacity at +20 °C.