High purity VOC regulators, Spectra Gas 7621
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21572 21572-R100
REST21572EA 3000 EUR
REST21572 REST21572-R100
High purity VOC regulators, Spectra Gas 7621
Gasregelaars
These single stage stainless steel VOC regulators feature stainless steel diaphragm, two pressure gauges, CGA-180 fitting and Kel-F® seat.
  • Accurate pressure control even at low flow rates
  • Individually tested for leaks and impurities