Inlet liners for PerkinElmer GCs
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20855 20733 20734 20743 20827 20828 20829 20830 20832 20833 20834 20835 20836 20853 20854 20899 20900 20910 20911 21717 21718 20738 20741 21026 21027 20832-209.1 20833-209 20833-211 20833-213 20833-214 20834-205 20834-213.25 20838-205 20840-214.5 20854-214 20900-210 20900-214 20911-214.5 21027-213.5 21718-205 21718-213
REST20855EA 0 EUR
REST20855 REST20733 REST20734 REST20743 REST20827 REST20828 REST20829 REST20830 REST20832 REST20833 REST20834 REST20835 REST20836 REST20853 REST20854 REST20899 REST20900 REST20910 REST20911 REST21717 REST21718 REST20738 REST20741 REST21026 REST21027 REST20832-209.1 REST20833-209 REST20833-211 REST20833-213 REST20833-214 REST20834-205 REST20834-213.25 REST20838-205 REST20840-214.5 REST20854-214 REST20900-210 REST20900-214 REST20911-214.5 REST21027-213.5 REST21718-205 REST21718-213
Inlet liners for PerkinElmer GCs
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
Use these liners for increased sensitivity.
  • They are manufactured from borosilicate glass or metal
  • Ø int. = Nominal diameter at syringe needle expulsion point