Static in-line mixers, HyperShear
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25341 25342 25343 26409 26410 26411 26413 26414
REST25341EA 1062 EUR
REST25341 REST25342 REST25343 REST26409 REST26410 REST26411 REST26413 REST26414
Static in-line mixers, HyperShear
Schudapparaten Vortexmengers
These stainless steel or PEEK in-line mixers are an efficient cross-flow shearing mechanism.
  • Reduced baseline noise
  • Increased sensitivity
  • Increased reaction efficiency in post-column derivatisation
  • Improved gradient accuracy for more reproducible results