HyClone™ Dialysed Foetal Bovine Serum, US origin
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30079.02 SH30079.03 SH30079.02HI SH30079.01HI SH30079.03HI SH30079.01 SH30079.03IR25
HYCLSH30079.02EA 166 EUR
HYCLSH30079.02 HYCLSH30079.03 HYCLSH30079.02HI HYCLSH30079.01HI HYCLSH30079.03HI HYCLSH30079.01 SH30079.03IR25
HyClone™ Dialysed Foetal Bovine Serum, US origin
Serum
Augment growth of cells with U.S.-sourced Cytiva HyClone Dialyzed Fetal Bovine Serum (FBS).
  • Depleted of small molecules (less than 10000 MW)
Filtered through sequential 100 nm (0.1 μm) pore-size rated filters.
Dialysed foetal bovine serum (FBS) is used in applications requiring serum depleted of small molecules (<10000 MW) and is filtered through filtered through three sequential 100 nm (0,1 µm) pore-size related filters. The diafiltration process reduces concentrations of low molecular weight components, such as nucleotides and amino acids, that are necessary for alternative biochemical survival pathways. The process is reproducible and reduces hypoxanthine and thymidine concentrations below detectable limits. The glucose concentration, which is representative of the extent of dialysis, is reported for each lot of serum.