Foetal bovine serum, research grade, South American origin, HyClone™
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SV30160.02 SV30160.03 SV30160.03HI SV30160.02HI
HYCLSV30160.02EA 68.3 EUR
HYCLSV30160.02 HYCLSV30160.03 HYCLSV30160.03HI HYCLSV30160.02HI
Foetal bovine serum, research grade, South American origin, HyClone™
Serum
HyClone™ research grade foetal bovine serum (FBS) collected and imported from South American countries in compliance with EU regulations (Directive 92/118/EEC) and has been filed as an in vitro diagnostic reagent at the AFSSAPS under registration number T86062. Strict manufacturing and quality control procedures ensure its quality and suitability for research applications. Research grade FBS is filtered through sequential 100 nm (0,1 µm) pore-size related filters.