FetalClone™ III serum, US origin
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30109.02 SH30109.03 SH30109.02HI SH30109.03HI SH30109.03IR25
SH30109.02EA 84.5 EUR
SH30109.02 HYCLSH30109.03 HYCLSH30109.02HI HYCLSH30109.03HI SH30109.03IR25
FetalClone™ III serum, US origin
Serum
Origin is the most widely applicable of the FetalClone family and is designed to include fibroblasts in its applications.
Conduct more cost-effective research without compromising performance. HyClone FetalClone III delivers a high culture yield at a reduced cost, plus it is readily available.