Thermolite® septa
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
27121 27122 27124 27127 27128 27130 27131 27134 27139 27140 27145 27146 27149
REST27121EA 0 EUR
REST27121 REST27122 REST27124 REST27127 REST27128 REST27130 REST27131 REST27134 REST27139 REST27140 REST27145 REST27146 REST27149
Thermolite® septa
Tussenschoten
These septa are preconditioned and ready to use.
  • Usable to 340 °C inlet temperature
  • Precision molding assures consistent, accurate fit
  • Excellent puncturability
Note: For 17 mm inlets, the maximum temperature for Thermolite septa is 290 °C.
Verpakking: Packaged in ultra-clean blister packs. 12,7 mm and 17 mm septa are packaged in precleaned glass jars.