Empty chromatography columns
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25118 25119 25120 25121 25122 25123 25124 25125 25126 25127 25128 25130 25131 25132 25133 25134 25135
REST25118EA 117 EUR
REST25118 REST25119 REST25120 REST25121 REST25122 REST25123 REST25124 REST25125 REST25126 REST25127 REST25128 REST25130 REST25131 REST25132 REST25133 REST25134 REST25135
Empty chromatography columns
Kolommen voor chromatografie
This 316 stainless steel tubings are complete with end-fittings, frits, nuts and ferrules which are preassembled. And the internal 1,6 mm seats are compatible with Valco® and parker fittings.