Whatman antibiotica assay discs
Leverancier: Whatman products (Cytiva)
Ratings:
Avg. Ratings: 4.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 4.0 out of 5
Was this review helpful? Yes No
Yes: 1 No: 1 
Distributeur de dispositifs médicaux / Directeur commercial / Cotonou
Yes, I recommend
Je voudrais vous demander la procédures pour acquérir les disques pour antibiogrammes. Je suis le directeur commercial d'une entreprise de distribution des équipements médicaux, produits chimiques et réactifs de laboratoire ainsi que des consommables médicaux. J'ai des clients qui désirent acquérir des disques pour antibiogrammes.
2017-006 2017-013 2017-009
710-0606EA 189 EUR
710-0606 710-0636 710-0635
Whatman antibiotica assay discs
Testsystemen voor microbiologie Antibioticatesten
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method. The antibiogram allows rational and selective chemotherapy. The test disks are coated with chemotherapeutic agents, placed on the inoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.
  • Uniform thick soft discs, for the absorption of test solutions in antibiotic assays
  • 440 g/m² and 920 µm
The antibiogram allows rational and selective chemotherapy. The test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.