Protein immunodetection reagents, SuperSignal™ West Femto maximum sensitivity substrate
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34095 34096
PIER34095EA 492 EUR
PIER34095 PIER34096
Protein immunodetection reagents, SuperSignal™ West Femto maximum sensitivity substrate
Reagentia voor elektroforese Western Blotting Reagents
SuperSignal™ West Femto maximum sensitivity substrate enables detection of low-femtogram amounts of protein deposited on nitrocellulose or PVDF membrane and probed with appropriate primary and secondary antibodies.
  • Highly sensitive with an intense signal which is easily captured on film or by imaging systems
  • Signal duration of 8 hours
  • Secondary antibodies included with the product
  • Can be stored at room temperature