Restore™ Western Blot Stripping Buffer
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21059 21062 21063
PIER21059EA 233 EUR
PIER21059 PIER21062 PIER21063
Restore™ Western Blot Stripping Buffer
Reagentia voor elektroforese Western Blotting Reagents
Although optimising assay conditions is the best way to achieve optimum results, re-performing the gel electrophoresis process to test each new primary antibody or antibody concentration is time-consuming and expensive. Restore™ Western blot stripping buffer offers an effective alternative.
  • Saves time - no need to re-run gels
  • Saves precious sample
  • Provides efficient removal
  • Gentle formulation ensures target protein is not damaged after stripping and re-probing
  • Odour-free