SuperSignal™ Protein ImmunoDetection Reagents, ELISA Femto Maximum Sensitivity Substrate
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
37075 37074
PIER37075EA 526 EUR
PIER37075 PIER37074
SuperSignal™ Protein ImmunoDetection Reagents, ELISA Femto Maximum Sensitivity Substrate
Reagentia voor elektroforese Western Blotting Reagents
SuperSignal® ELISA Femto maximum sensitivity substrate utilises the SuperSignal® system with an enhancer formulation for superior protein detection and low-end linearity in ELISA applications. Ideal for automated high-throughput screening applications.
  • Intense signal generated immediately at both room temperature and 37 °C
  • Easy detection of low quantities of proteins with high signal:noise ratios and low-end linearity of dose response curves
  • Femtogram-level detection of target proteins in ELISA
  • High sensitivity allows for reduction in ELISA incubation steps and hence a reduction in assay time
  • Storage at room temperature with a 6 hour working solution stability