HPLC column tubing
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25100 25101 25102 25103 25104 25105 25106 25107 25108 25109 25110 25111 25112 25113 25114 25115 25116 25117
REST25100EA 52 EUR
REST25100 REST25101 REST25102 REST25103 REST25104 REST25105 REST25106 REST25107 REST25108 REST25109 REST25110 REST25111 REST25112 REST25113 REST25114 REST25115 REST25116 REST25117
HPLC column tubing
Slangen Chromatography Tubing
Manufactured from fine, chromatographic grade 316 stainless steel.
  • Pre polished tubing ensures burr-free cut
  • No additional treatment needed
Corrosion resistant, ultrasonically cleaned, and passivated.