Packed column inlet adapter kits
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21650 21651
REST21650EA 57 EUR
REST21650 REST21651
Packed column inlet adapter kits
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Packed column inlet adapter kits center columns perfectly in 6,35 mm injection ports to eliminate bent syringe needles.
  • Slotted design prevents carrier gas occlusion
  • Vespel/graphite-reducing ferrules make installation easy
  • Centers column perfectly in injection port to eliminate bent syringe needles
  • Adapter kits include all nuts and ferrules used to attach tubing to the injector or detector