HPLC frits
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25074 25071 25072 25073 25075 25076
REST25074EA 158 EUR
REST25074 REST25071 REST25072 REST25073 REST25075 REST25076
HPLC frits
Filters Voorkolom filters en frits
These 6 mm frits are manufactured from fine, chromatographic grade 316 stainless steel and different sizes are available to fit most column and pore sizes. Compatible frit have to be chosen by checking its pore size with particle size of the packaging in the column.