Labels for BBP33 printers, type B-432, ToughBond™
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
143003 143004 143054 133803
BRDY143003EA 521.31 EUR
BRDY143003 BRDY143004 BRDY143054 BRDY133803
Labels for BBP33 printers, type B-432, ToughBond™
Etiketten
These polyester labels have a gloss finish and are suitable for labeling on smooth and textured surfaces.
  • Good moisture resistance
  • High adhesion
Used for product and general purpose identification, bin and shelf identification, barcoding and asset tracking, laboratory identification.