Scrapers, LaboPlast® / SteriPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5378-0030 5378-1030
BURK5378-0030EA 19.6 EUR
BURK5378-0030 BURK5378-1030
Scrapers, LaboPlast® / SteriPlast®
Schrapers
PS, white. Sturdy and convenient scrapers for disposable use. The scrapers are suitable for scraping off, smoothing or cleaning as well as removing residue.
  • Produced and packaged in a class 7 cleanroom
  • Suitable for food contact
  • Individually packaged and optionally sterilised by gamma rays