Spoon spatulas, LaboPlast® / SteriPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5378-0031 5378-1031
231-0608EA 23.6 EUR
231-0608 231-0609
Spoon spatulas, LaboPlast® / SteriPlast®
Spatels
PS. Double spatula with a 0,5 ml spoon and a 17 mm spatula blade. Ideal for use as a sampling spoon, for measuring or transferring small amounts, as a scraper or for mixing and stirring.
  • Resistant to chemicals, corrosion-proof
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
  • Optionally packaged individually and sterilised by gamma rays
Verpakking: Available either in a large package with 100 pieces or 100 pieces packaged individually and sterilised by gamma rays.