Capping A 20% (v/v) n-Methylimidazole in ACN, Novabiochem® for DNA/RNA synthesis
Leverancier: Novabiochem (Part of Merck)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8.57002.0500 8.57002.1000
BI0224/0505EA 0 EUR
BI0224/0505 BI0224/1005
Capping A 20% (v/v) n-Methylimidazole in ACN, Novabiochem® for DNA/RNA synthesis
Capping Reagent
Capping reagent A for DNA/RNA synthesis high quality, lowest water and acid content are best requirements for a reliable and reproducible DNA-/RNA synthesis with high yields.
  • 20% (v/v) n-Methylimidazole in Acetonitrile
  • Packed in glass bottle
  • Storage temperature below 30 °C