Aflatoxin Mix, analytical standards for chromatography, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34036-1ML-R 34036-2ML-R
34036-1ML-REA 347 EUR
34036-1ML-R 34036-2ML-R
Aflatoxin Mix, analytical standards for chromatography, Supelco®
Standaarden Organische standaarden
These analytical standards are suitable for HPLC and gas chromatography. Mutil component solution with a concentration of 0,5 μg/ml B2 and G2 in acetonitrile and 2 μg/ml B1 and G1 in acetonitrile.