Valves for HPLC
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26407 26400 26401 26402 26403 26404 26406
REST26407EA 68 EUR
REST26407 REST26400 REST26401 REST26402 REST26403 REST26404 REST26406
Valves for HPLC
Ventielen Terugslagventielen
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
  • Only fingertight torque is required to make the seal