Back pressure regulators
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25017 25018 25020
REST25017EA 311 EUR
REST25017 REST25018 REST25020
Back pressure regulators
Ventielen Terugslagventielen
Backpressure regulators can improve detector performance by preventing bubble formation in the detector flow cell. They also are useful in post-column reaction lines and between detectors and fraction collectors in preparatory work. Regulators are superior to more specific alternative solutions like small bore tubing, in which pressure varies with flow rate.
Both types are adjustable to ensure constant backpressure over a wide range of mobile phase viscosities and flow rates. The end-of-line model is available with ¹/₄ to 28 plastic flange-type fittings or high-pressure 1,6 mm compressionMore Product Information