Column connectors and plugs
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25015 25016
REST25015EA 69 EUR
REST25015 REST25016
Column connectors and plugs
Fittingen en connectoren
Universal 10-32 PEEK connectors for easy installation of all ¹/₁₆" tubing, it includes stainless steel, up to 345 bar.