Waste overflow indicator for HPLC systems
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26543 26550
REST26543EA 378 EUR
REST26543 REST26550
Waste overflow indicator for HPLC systems
Niveausensoren
The waste overflow indicator keeps the mobile phase waste in the waste container.
  • Avoid messy pooling around mobile phase waste containers
  • Audible alarm instantly alerts user, preventing overflow
  • Compact, battery operated unit
Compact, battery operated unit fits securely on solvent bottles and accommodates two waste streams. An audible alarm is given as the solvent waste container approaches capacity, giving you time to empty or change the container.