Chemisch bestendige pak, Microchem® by AlphaTec®, model 151-G02
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GR40T-00151-03 GR40T-00151-04 GR40T-00151-05 GR40T-00151-06 GR40T-00151-07
ANSEGR40T-00151-03EA 1160 EUR
ANSEGR40T-00151-03 ANSEGR40T-00151-04 ANSEGR40T-00151-05 ANSEGR40T-00151-06 ANSEGR40T-00151-07
Chemisch bestendige pak, Microchem® by AlphaTec®, model 151-G02
Wegwerpkleding
Rear entry suit with neoprene rubber face seal for a close fit to full face respirators. Ideal for use in hazardous areas where protection against concentrated chemicals and biological agents is required.
  • Neoprene rubber face seal
  • Rear horizontal zip entry
  • Attached socks with boot overflap
  • Attached Ansell Barrier gloves, with oversleeves and finger loops
  • Ultrasonically welded and taped seams provide a high barrier to liquid and particulates
Typical applications include: Chemicals, oils and petrochemicals, pharmaceutical, agriculture, sewage purification installations, industrial and tank cleaning and emergency services (HAZMAT, CBRN). Suitable for Ex-Zones.
Certificaten: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III