ESD safety ankle boots, lace-up, Jupiter, Jalas® 1828
Leverancier: EJENDALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1828-42 1828-44 1828-47 1828-35 1828-36 1828-37 1828-38 1828-39 1828-40 1828-41 1828-43 1828-45 1828-46 1828-48 1828-49 1828-50
EJEN1828-42EA 129.5 EUR
EJEN1828-42 EJEN1828-44 EJEN1828-47 EJEN1828-35 EJEN1828-36 EJEN1828-37 EJEN1828-38 EJEN1828-39 EJEN1828-40 EJEN1828-41 EJEN1828-43 EJEN1828-45 EJEN1828-46 EJEN1828-48 EJEN1828-49 EJEN1828-50
ESD safety ankle boots, lace-up, Jupiter, Jalas® 1828
Schoenen en laarzen
Wide fit, anti-static ESD safety ankle boots made from PU coated leather with EVA, polyester-based conductive thread insole, polyester/polyamide lining plus a heat and oil resistant outsole.
  • Double shock-absorption zones in Poron® XRD®
  • ProNose toe reinforcement
  • Puncture resistant steel midsole
  • Toe protection cap made from aluminium (200 J)
Mainly used for outdoors, environments with risk for outsole penetration and harsh environments.
Colour: Black/grey/yellow
Certificaten: EN 61340-5-1 (ESD); EN ISO 20345:2011; S3, SRC, CI, HRO