Wash kit for RHS or other PAL/CTC GC autosamplers, Chromacol™
Leverancier: CHROMACOL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WSKT-25 WSKTCL-100
CRMAWSKT-25EA 75.4 EUR
CRMAWSKT-25 CRMAWSKTCL-100
Wash kit for RHS or other PAL/CTC GC autosamplers, Chromacol™
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
This wash kit with 25 vials and caps is suitable for equipping a lab with 4 or 5 instruments or one instrument for a longer period. The value of regularly replacing caps is not always clear so many labs do not routinely maintain the wash stations.
  • Improved sealing, less evaporation of wash solvent
  • Easy to apply caps
  • Elimination of wash solvent contamination
Wash vials on GC autosamplers are commonly left open or have caps that are difficult to apply. Without the caps there is a risk of the solvent being lost and the syringe wash not being correctly carried out. Contamination from the wash solvent is also possible once the injections are carried out due to septa being carried into the solvent.