ESD safety shoes, lace-up, E-Sport, JALAS® 1615
Leverancier: EJENDALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1615-36 1615-37 1615-38 1615-39 1615-40 1615-41 1615-42 1615-43 1615-44 1615-45 1615-46 1615-47
EJEN1615-36EA 83.27 EUR
EJEN1615-36 EJEN1615-37 EJEN1615-38 EJEN1615-39 EJEN1615-40 EJEN1615-41 EJEN1615-42 EJEN1615-43 EJEN1615-44 EJEN1615-45 EJEN1615-46 EJEN1615-47
ESD safety shoes, lace-up, E-Sport, JALAS® 1615
Schoenen en laarzen
Anti-static, lightweight, ESD safety shoes made from PU coated split leather with EVA, polyester-based conductive thread insole and polyester/polyamide lining.
  • Shock-absorption zone in Ergothan
  • Plastic shank insole and PU outsole
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
  • Toe protection cap made from aluminium (200 J)
Mainly used for outdoors, indoors and environments with risk for outsole penetration.
Colour: Black/grey/red
Certificaten: EN 61340-5-1 (ESD); EN ISO 20345:2011; S3, SRC, CI