ESD safety low shoes, Zenit Easyroll, JALAS® 1738
Leverancier: EJENDALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1738-42 1738-36 1738-37 1738-38 1738-39 1738-40 1738-41 1738-43 1738-44 1738-45 1738-46 1738-47
EJEN1738-42EA 156.98 EUR
EJEN1738-42 EJEN1738-36 EJEN1738-37 EJEN1738-38 EJEN1738-39 EJEN1738-40 EJEN1738-41 EJEN1738-43 EJEN1738-44 EJEN1738-45 EJEN1738-46 EJEN1738-47
ESD safety low shoes, Zenit Easyroll, JALAS® 1738
Schoenen en laarzen
Anti-static ESD safety shoes made from PU coated split leather with EVA, polyester-based conductive thread insole and merino wool lining.
  • Double shock-absorption zones in Poron® XRD
  • Boa® fastening
  • ProNose toe reinforcement
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
  • Toe protection cap made from aluminium (200 J)
Mainly used for outdoors, indoors and environments with risk for outsole penetration.
Colour: Black/grey/red
Certificaten: EN 61340-5-1 (ESD); EN ISO 20345:2011; S3 SRC