Schort met mouwen, AlphaTec® 4000, model 215
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GR40T-00215-05 GR40T-00215-06 GR40T-00215-03 GR40T-00215-04 GR40T-00215-02 GR40T-00215-07
113-9968EA 1070 EUR
113-9968 113-9969 113-9966 113-9967 113-9965 113-9970
Schort met mouwen, AlphaTec® 4000, model 215
Wegwerpkleding
This apron is made from a unique multi-layer barrier fabric (External film barrier layer, non woven inner layer, core high barrier film layer) renowned for its lightweight, yet robust textile feel and exceptional barrier to organic and inorganic chemicals.
  • Lange mouwen
  • Klittenbandsluiting aan de nek
  • Lintjessluiting aan de taille
  • Dubbele manchet met gebreide binnenmanchet
  • Ultrasoon gelaste en getapete naden
Recommended for: Contaminated environments; chemical handling; filling, blending and charging of raw materials; transferring liquids and solids; opening and draining pumps valves or lines; raw material sample collection; concentration mixing and palletisation; Hazmat response; assembly and disassembly of large pipes.
Certificaten: Type PB (3)/PB (4); EN 14126 (PB(3)-B/PB (4)-B); Cat. III