Test kits in ampoules, AQUAfast™ IV reagents, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AC4004 AC4005 AC4006 AC4007 AC4011 AC4012 AC4016 AC4017 AC4046 AC4048 AC4055 AC4060 AC4065 AC4070 AC4078 AC4082 AC4095 AC4099
ORIOAC4004EA 243 EUR
ORIOAC4004 ORIOAC4005 ORIOAC4006 705-0751 ORIOAC4011 ORIOAC4012 ORIOAC4016 ORIOAC4017 ORIOAC4046 ORIOAC4048 ORIOAC4055 ORIOAC4060 ORIOAC4065 705-0752 ORIOAC4078 ORIOAC4082 705-0753 ORIOAC4099
Test kits in ampoules, AQUAfast™ IV reagents, Orion™
Milieutestkits
Orion™ AQUAfast™ Auto-Test Ampoule Reagents can be used with the Orion™ AQUAfast™ AQ4000 colorimeter to simplify sample preparation and tests.
  • Consistency and reproducibility improved with AQUAfast™ ampoules and an AQ4000 meter
  • Auto-ID™ eliminates measurement errors
  • Designed for use with colorimetric water and wastewater testing equipment
The Auto-Test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.