Solvent debubbler
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
REST25014EA 101 EUR
REST25014
Solvent debubbler
Ontgassers
Bubbles in an LC system can cause check valve malfunctions and pump cavitation, seriously affecting pump performance. The debubbler removes bubbles from the fluid stream before it enters the pump.
  • The gas/liquid interface is easily visible through the translucent wall
  • Loosening the airtight cap releases the trapped gas
The debubbler features a PP body and bracket; universal inlet and outlet connecting tips are made of 316 stainless steel and accept 0,063", 0,085", and 0,125" ID tubing.