Beakers, quartz, tall form, with spout
Leverancier: TECHNICAL QUARTZ SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75230 75231 75222 75223 75224 75225 75226 75227 75228 75229
TSLA75230EA 24.2 EUR
TSLA75230 TSLA75231 TSLA75222 TSLA75223 TSLA75224 TSLA75225 TSLA75226 TSLA75227 TSLA75228 TSLA75229
Beakers, quartz, tall form, with spout
Bekers
Quartz glass, with spout.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensable for high-technology applications.