Sterlisation gusseted pouches, Stericlin®
Leverancier: VP Medical Packaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3FKSB220108 3FKSB220114 3FKSB220102 3FKSB220104 3FKSB220106 3FKSB220110
129-1266EA 481 EUR
129-1266 129-1269 129-1263 129-1264 129-1265 129-1267
Sterlisation gusseted pouches, Stericlin®
Zakken Sterilisatiezakken
These see-through sterilisation pouches made of sterilisation proof 70 g/m² heavy-duty paper are a universal packaging solution.
  • Effective barrier against germs and bacteria
  • Permits the transmission of air, steam or sterilisation gases
  • Three indicators for STEAM-, EO- and FORM-sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Certificaten: DIN EN ISO 11607 and DIN 868-5.