Anion exchange chromatography column kits, NucleoBond® Xtra Midi/Maxi
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
740416.10 740410.50 740416.50 740410.10 740410.100 740412.10 740412.50 740414.10 740414.100 740414.50
MANA740416.10EA 280 EUR
MANA740416.10 MANA740410.50 MANA740416.50 MANA740410.10 MANA740410.100 MANA740412.10 MANA740412.50 MANA740414.10 MANA740414.100 MANA740414.50
Anion exchange chromatography column kits, NucleoBond® Xtra Midi/Maxi
Kolommen voor chromatografie
NucleoBond® Xtra Midi and Maxi kits contain enlarged columns which leads to lower silica resin beds. This in turn enables faster flow of lysate and buffers through the columns.
Specially designed column filters are included for convenient and time-saving clarification of bacterial lysates. The column filters are supplied inserted in the NucleoBond® Xtra columns and allow parallel clarification of bacterial lysate and loading onto the column.
Leveringsinformatie: NucleoBond® Xtra Midi and Maxi columns supplied with inserted NucleoBond® Xtra column Filters, buffers, RNase A (10, 50 and 100 preps), NucleoBond® Xtra Midi Plus and Maxi Plus columns provided with NucleoBond® Finalizers in additional (10 and 50 preps).