Drums, wide mouth, UN approved
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1361 1363 1364 1353 1351 1352
LAMA1361EA 127 EUR
LAMA1361 LAMA1363 LAMA1364 LAMA1353 LAMA1351 LAMA1352
Drums, wide mouth, UN approved
Vaten, vaten, trommels
Drums, made of HDPE, blue, UN approved, with lid.
  • Stackable
  • Tamper evident
  • Self-sealing lid with zinc plate steel closure
  • For food use
For food, cosmetic and chemical fields. Manufactured with the inner neutral layer that avoids the contact between the content and the coloring pigment.
Dimensions allow overlap on pallet 800×1200 and 1000×1200.