Production and laboratory chairs with synchronous technology, Neon 3
Leverancier: bimos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2003280525807 2003279525807 2003278525807 SP13280525807
132-1447EA 774 EUR
132-1447 132-1446 132-1445 132-1451
Production and laboratory chairs with synchronous technology, Neon 3
Stoelen
Laboratory chair neon with closed mechanism and synchronous technology.
  • Black upholstery superfabric
  • Non slip with seat inclination adjustment
  • Seat height adjustment with a gasspring from 590 to 870 mm
  • 5-star base aluminium stop-and-go-castors, with foot ring